Learn More
Biomatik Corporation Recombinant Danio rerio Superoxide dismutase [Cu-Zn] (sod1), 100UG
Supplier:  Biomatik Corporation RPC20493
Encompass_Preferred
Recombinant Danio rerio Superoxide dismutase [Cu-Zn] (sod1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Danio rerio (Zebrafish) (Brachydanio rerio). Target Name: sod1. Target Synonyms: sod1; cuzn; Superoxide dismutase [Cu-Zn]; EC 1.15.1.1. Accession Number: O73872. Expression Region: 1~154aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 37.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4840