Recombinant Human Forkhead box protein M1 (FOxM1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: FOXM1. Target Synonyms: Forkhead-related protein FKHL16Hepatocyte nuclear factor 3 forkhead homolog 11. Accession Number: Q08050. Expression Region: 235~327aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 31.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.