Learn More
Biomatik Corporation Recombinant Atlantic salmon Vertebrate ancient opsin, partial, 100UG
Catalog No. 501504845
Encompass_Preferred
Recombinant Atlantic salmon Vertebrate ancient opsin is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Salmo salar (Atlantic salmon). Target Name: Atlantic salmon Vertebrate ancient opsin. Target Synonyms: Vertebrate ancient opsin. Accession Number: O13018. Expression Region: 1~75aa. Tag Info: N-Terminal 6Xhis-B2M-Tagged. Theoretical MW: 22.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4845 Supplier Biomatik Corporation Supplier No. RPC20498
Recombinant Atlantic salmon Vertebrate ancient opsin is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Salmo salar (Atlantic salmon). Target Name: Atlantic salmon Vertebrate ancient opsin. Target Synonyms: Vertebrate ancient opsin. Accession Number: O13018. Expression Region: 1~75aa. Tag Info: N-Terminal 6Xhis-B2M-Tagged. Theoretical MW: 22.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.