Learn More
Biomatik Corporation Recombinant Human ATP synthase subunit beta, mitochondrial (ATP5B), 100UG
Supplier:  Biomatik Corporation RPC20504
Encompass_Preferred
Recombinant Human ATP synthase subunit beta, mitochondrial (ATP5F1B) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ATP5F1B. Target Synonyms: ATP 5B; ATP synthase H+ transporting mitochondrial F1 complex beta polypeptide; ATP synthase subunit beta mitochondrial; ATP synthase subunit beta; mitochondrial; atp5b; ATPB; ATPB_HUMAN; ATPMB; ATPSB; Epididymis secretory protein Li 271. Accession Number: P06576. Expression Region: 48~529aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 53.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4849