Learn More
Biomatik Corporation Recombinant Escherichia coli Chaperone protein HtpG (htpG), 100UG
Supplier:  Biomatik Corporation RPC20508
Encompass_Preferred
Recombinant Escherichia coli Chaperone protein HtpG (htpG) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia coli (strain K12). Target Name: htpG. Target Synonyms: Heat shock protein C62.5Heat shock protein HtpGHigh temperature protein G. Accession Number: P0A6Z3. Expression Region: 1~624aa. Tag Info: Tag-Free. Theoretical MW: 71.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4852