Learn More
Biomatik Corporation Recombinant Escherichia coli Putative binding protein ygiS (ygiS), 100UG
Supplier:  Biomatik Corporation RPC20515
Encompass_Preferred
Recombinant Escherichia coli Putative binding protein ygiS (ygiS) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia coli (strain K12). Target Name: ygiS. Target Synonyms: ygiS; b3020; JW2988; Probable deoxycholate-binding periplasmic protein YgiS. Accession Number: Q46863. Expression Region: 21~535aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 74.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4859