Learn More
Biomatik Corporation Recombinant Human Stromelysin-2 (MMP10), 100UG
Supplier:  Biomatik Corporation RPC20061
Encompass_Preferred
Recombinant Human Stromelysin-2 (MMP10) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: MMP10. Target Synonyms: Matrix metalloproteinase-10; MMP-10Transin-2. Accession Number: P09238. Expression Region: 99~476aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 47kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4563