Learn More
Biomatik Corporation Recombinant Escherichia coli Periplasmic murein peptide-binding protein (mppA), 100UG
Supplier:  Biomatik Corporation RPC20517
Encompass_Preferred
Recombinant Escherichia coli Periplasmic murein peptide-binding protein (mppA) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia coli (strain K12). Target Name: mppA. Target Synonyms: mppA; ynaH; b1329; JW1322; Periplasmic murein peptide-binding protein. Accession Number: P77348. Expression Region: 23~537aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 73.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4861