Learn More
Biomatik Corporation Recombinant Mouse Tissue alpha-L-fucosidase (Fuca1), 100UG
Supplier:  Biomatik Corporation RPC20521
Encompass_Preferred
Recombinant Mouse Tissue alpha-L-fucosidase (Fuca1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Fuca1. Target Synonyms: Alpha-L-fucosidase IAlpha-L-fucoside fucohydrolase 1. Accession Number: Q99LJ1. Expression Region: 18~452aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 66.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4863