Learn More
Biomatik Corporation Recombinant Streptomyces clavuligerus Beta-lactamase inhibitory protein, 100UG
Catalog No. 501504866
Encompass_Preferred
Recombinant Streptomyces clavuligerus Beta-lactamase inhibitory protein is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Streptomyces clavuligerus. Target Name: Streptomyces clavuligerus Beta-lactamase inhibitory protein. Target Synonyms: Beta-lactamase inhibitory protein; BLIP. Accession Number: P35804. Expression Region: 37~201aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 33.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4866 Supplier Biomatik Corporation Supplier No. RPC20524
Recombinant Streptomyces clavuligerus Beta-lactamase inhibitory protein is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Streptomyces clavuligerus. Target Name: Streptomyces clavuligerus Beta-lactamase inhibitory protein. Target Synonyms: Beta-lactamase inhibitory protein; BLIP. Accession Number: P35804. Expression Region: 37~201aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 33.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.