Learn More
Biomatik Corporation Recombinant Human Inositol hexakisphosphate kinase 1 (IP6K1), 100UG
Supplier:  Biomatik Corporation RPC20525
Encompass_Preferred
Recombinant Human Inositol hexakisphosphate kinase 1 (IP6K1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: IP6K1. Target Synonyms: Inositol hexaphosphate kinase 1. Accession Number: Q92551. Expression Region: 1~441aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 66.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4867