Learn More
Biomatik Corporation Recombinant Rat FAD-linked sulfhydryl oxidase ALR (Gfer), 100UG
Catalog No. 501504868
Encompass_Preferred
Recombinant Rat FAD-linked sulfhydryl oxidase ALR (Gfer) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rat (Rattus norvegicus). Target Name: Gfer. Target Synonyms: Augmenter of liver regeneration. Accession Number: Q63042. Expression Region: 1~198aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 38.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4868 Supplier Biomatik Corporation Supplier No. RPC20526
Recombinant Rat FAD-linked sulfhydryl oxidase ALR (Gfer) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rat (Rattus norvegicus). Target Name: Gfer. Target Synonyms: Augmenter of liver regeneration. Accession Number: Q63042. Expression Region: 1~198aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 38.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.