Learn More
Biomatik Corporation Recombinant Neisseria meningitidis serogroup B Penicillin-binding protein 1A (mrcA), partial, 100UG
Supplier:  Biomatik Corporation RPC20531
Encompass_Preferred
Recombinant Neisseria meningitidis serogroup B Penicillin-binding protein 1A (mrcA), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Neisseria meningitidis serogroup B (strain MC58). Target Name: mrcA. Target Synonyms: Peptidoglycan TGaseDD-transpeptidase. Accession Number: P0A0Z6. Expression Region: 206~413aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 40kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4873