Learn More
Biomatik Corporation Recombinant Human herpesvirus 2 Envelope glycoprotein B (gB), partial, 100UG
Supplier:  Biomatik Corporation RPC20534
Encompass_Preferred
Recombinant Human herpesvirus 2 Envelope glycoprotein B (gB), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human herpesvirus 2 (strain HG52) (HHV-2) (Human herpes simplex virus 2). Target Name: gB. Target Synonyms: gB; UL27; Envelope glycoprotein B; gB. Accession Number: P08666. Expression Region: 23~279aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 44.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4876