Learn More
Biomatik Corporation Recombinant Dahlia merckii Defensin-like protein 1, 100UG
Catalog No. 501504877
Encompass_Preferred
Recombinant Dahlia merckii Defensin-like protein 1 is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Dahlia merckii (Bedding dahlia). Target Name: Dahlia merckii Defensin-like protein 1. Target Synonyms: Cysteine-rich antimicrobial protein 1Defensin AMP1. Accession Number: P0C8Y4. Expression Region: 1~50aa. Tag Info: N-Terminal 6Xhis-B2M-Tagged. Theoretical MW: 19.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4877 Supplier Biomatik Corporation Supplier No. RPC20535
Recombinant Dahlia merckii Defensin-like protein 1 is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Dahlia merckii (Bedding dahlia). Target Name: Dahlia merckii Defensin-like protein 1. Target Synonyms: Cysteine-rich antimicrobial protein 1Defensin AMP1. Accession Number: P0C8Y4. Expression Region: 1~50aa. Tag Info: N-Terminal 6Xhis-B2M-Tagged. Theoretical MW: 19.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.