Learn More
Biomatik Corporation Recombinant Rat Frataxin, mitochondrial (Fxn), 100UG
Catalog No. 501504879
Encompass_Preferred
Recombinant Rat Frataxin, mitochondrial (Fxn) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rat (Rattus norvegicus). Target Name: Fxn. Target Synonyms: FxnFrataxin; mitochondrial; Fxn; EC 1.16.3.1; Cleaved into: Frataxin intermediate form; Frataxin mature form. Accession Number: D3ZYW7. Expression Region: 41~208aa. Tag Info: Tag-Free. Theoretical MW: 18.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4879 Supplier Biomatik Corporation Supplier No. RPC20538
Recombinant Rat Frataxin, mitochondrial (Fxn) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rat (Rattus norvegicus). Target Name: Fxn. Target Synonyms: FxnFrataxin; mitochondrial; Fxn; EC 1.16.3.1; Cleaved into: Frataxin intermediate form; Frataxin mature form. Accession Number: D3ZYW7. Expression Region: 41~208aa. Tag Info: Tag-Free. Theoretical MW: 18.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.