Learn More
Biomatik Corporation Recombinant Human Bifunctional heparan sulfate N-deacetylase/N-sulfotransferase 1 (NDST1), partial, 100UG
Supplier:  Biomatik Corporation RPC20063
Encompass_Preferred
Recombinant Human Bifunctional heparan sulfate N-deacetylase/N-sulfotransferase 1 (NDST1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: NDST1. Target Synonyms: Glucosaminyl N-deacetylase/N-sulfotransferase 1; NDST-1N-heparan sulfate sulfotransferase 1; N-HSST 1[Heparan sulfate]-glucosamine N-sulfotransferase 1. Accession Number: P52848. Expression Region: 40~556aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 73.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4565