Learn More
Biomatik Corporation Recombinant Human Kallikrein-2 (KLK2), 100UG
Catalog No. 501504881
Encompass_Preferred
Recombinant Human Kallikrein-2 (KLK2) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: KLK2. Target Synonyms: Glandular kallikrein-1. Accession Number: P20151. Expression Region: 25~261aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 30.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4881 Supplier Biomatik Corporation Supplier No. RPC20540
Recombinant Human Kallikrein-2 (KLK2) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: KLK2. Target Synonyms: Glandular kallikrein-1. Accession Number: P20151. Expression Region: 25~261aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 30.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.