Learn More
Biomatik Corporation Recombinant Chlamydomonas reinhardtii Autolysin, 100UG
Supplier:  Biomatik Corporation RPC20541
Encompass_Preferred
Recombinant Chlamydomonas reinhardtii Autolysin is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Chlamydomonas reinhardtii (Chlamydomonas smithii). Target Name: Chlamydomonas reinhardtii Autolysin. Target Synonyms: Gamete lytic enzyme. Accession Number: P31178. Expression Region: 184~638aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 65.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4882