Learn More
Biomatik Corporation Recombinant Wuchereria bancrofti Putative serine protease inhibitor, 100UG
Supplier:  Biomatik Corporation RPC20545
Encompass_Preferred
Recombinant Wuchereria bancrofti Putative serine protease inhibitor is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Wuchereria bancrofti. Target Name: Wuchereria bancrofti Putative serine protease inhibitor. Accession Number: F6KME1. Expression Region: 1~371aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 62.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4886