Learn More
Biomatik Corporation Recombinant Mycobacterium tuberculosis Putative amidase AmiB2 (amiB2), 100UG
Supplier:  Biomatik Corporation RPC20549
Encompass_Preferred
Recombinant Mycobacterium tuberculosis Putative amidase AmiB2 (amiB2) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mycobacterium tuberculosis (strain ATCC 25618 / H37Rv). Target Name: amiB2. Target Synonyms: amiB2; Rv1263; MTCY50.19cPutative amidase AmiB2; EC 3.5.1.4. Accession Number: P9WQ97. Expression Region: 1~462aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 69.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4889