Learn More
Biomatik Corporation Recombinant Escherichia coli K99 fimbrial protein (fanC), 100UG
Catalog No. 501504896
Encompass_Preferred
Recombinant Escherichia coli K99 fimbrial protein (fanC) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia coli. Target Name: fanC. Target Synonyms: fanCK99 fimbrial protein. Accession Number: P18103. Expression Region: 23~181aa. Tag Info: Tag-Free. Theoretical MW: 16.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4896 Supplier Biomatik Corporation Supplier No. RPC20557
Recombinant Escherichia coli K99 fimbrial protein (fanC) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia coli. Target Name: fanC. Target Synonyms: fanCK99 fimbrial protein. Accession Number: P18103. Expression Region: 23~181aa. Tag Info: Tag-Free. Theoretical MW: 16.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.