Learn More
Biomatik Corporation Recombinant Human HNRNPA2B1 protein (HNRNPA2B1), 100UG
Catalog No. 501504905
Encompass_Preferred
Recombinant Human Heterogeneous nuclear ribonucleoproteins A2/B1 (HNRNPA2B1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: HNRNPA2B1. Accession Number: I6L957. Expression Region: 1~249aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 55.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4905 Supplier Biomatik Corporation Supplier No. RPC20574
Recombinant Human Heterogeneous nuclear ribonucleoproteins A2/B1 (HNRNPA2B1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: HNRNPA2B1. Accession Number: I6L957. Expression Region: 1~249aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 55.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.