Learn More
Biomatik Corporation Recombinant Human Prostaglandin-H2 D-isomerase (PTGDS), 100UG
Supplier:  Biomatik Corporation RPC20067
Encompass_Preferred
Recombinant Human Prostaglandin-H2 D-isomerase (PTGDS) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: PTGDS. Target Synonyms: Beta-trace protein; Cerebrin-28Glutathione-independent PGD synthaseLipocalin-type prostaglandin-D synthaseProstaglandin-D2 synthase; PGD2 synthase; PGDS; PGDS2. Accession Number: P41222. Expression Region: 23~190aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 34.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4568