Learn More
Biomatik Corporation Recombinant Human HLA class I histocompatibility antigen, alpha chain G protein (HLA-G), 100UG
Supplier:  Biomatik Corporation RPC20584
Encompass_Preferred
Recombinant Human HLA class I histocompatibility antigen, alpha chain G (HLA-G) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Mammalian Cell. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: HLA-G. Target Synonyms: HLA G antigen; MHC class I antigen G. Accession Number: P17693. Expression Region: 25~338aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 39.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4910