Learn More
Biomatik Corporation Recombinant Human Zyxin (ZYX), 100UG
Supplier:  Biomatik Corporation RPC20585
Encompass_Preferred
Recombinant Human Zyxin (ZYX) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Mammalian Cell. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ZYX. Target Synonyms: Zyxin-2. Accession Number: Q15942. Expression Region: 2~572aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 65.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4911