Learn More
Biomatik Corporation Recombinant Human Pyridoxal phosphate phosphatase (PDXP), 100UG
Supplier:  Biomatik Corporation RPC20594
Encompass_Preferred
Recombinant Human Pyridoxal phosphate phosphatase (PDXP) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Baculovirus. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: PDXP. Target Synonyms: PLP phosphataseChronophin. Accession Number: Q96GD0. Expression Region: 1~296aa. Tag Info: C-Terminal 9Xhis-Tagged. Theoretical MW: 33.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4916