Learn More
Biomatik Corporation Recombinant Pig Heat shock protein HSP 90-alpha (HSP90AA1), partial, 100UG
Supplier:  Biomatik Corporation RPC20596
Encompass_Preferred
Recombinant Pig Heat shock protein HSP 90-alpha (HSP90AA1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Baculovirus. Endotoxin Level: Not Tested. Species: Pig (Sus scrofa; Porcine). Target Name: HSP90AA1. Target Synonyms: HSP90AA1; HSP90A; HSPCAHeat shock protein HSP 90-alpha. Accession Number: O02705. Expression Region: 222~367aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 19.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4917