Learn More
Biomatik Corporation Recombinant Mouse Glutaminyl-peptide cyclotransferase (Qpct), 100UG
Supplier:  Biomatik Corporation RPC20598
Encompass_Preferred
Recombinant Mouse Glutaminyl-peptide cyclotransferase (Qpct) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Mammalian Cell. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Qpct. Target Synonyms: Glutaminyl cyclaseQC. Accession Number: Q9CYK2. Expression Region: 36~362aa. Tag Info: N-Terminal 6Xhis-Myc-Tagged. Theoretical MW: 41.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4918