Learn More
Biomatik Corporation Recombinant Salmonella enterica OmpA family protein (A673_03341), partial, 100UG
Supplier:  Biomatik Corporation RPC20600
Encompass_Preferred
Recombinant Salmonella enterica OmpA family protein (A673_03341), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Mammalian Cell. Endotoxin Level: Not Tested. Species: Salmonella enterica subsp. enterica serovar Enteritidis str. 2009K0958. Target Name: A673_03341. Accession Number: S4JJH7. Expression Region: 82~220aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 18.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4920