Learn More
Biomatik Corporation Recombinant Human Folate receptor beta (FOLR2), partial, 100UG
Supplier:  Biomatik Corporation RPC20627
Encompass_Preferred
Recombinant Human Folate receptor beta (FOLR2), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Baculovirus. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: FOLR2. Target Synonyms: Folate receptor 2Folate receptor, fetal/placentalPlacental folate-binding protein. Accession Number: P14207. Expression Region: 19~230aa. Tag Info: N-Terminal 10Xhis-Tagged. Theoretical MW: 27.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4925