Recombinant Human Optineurin (OPTN) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Baculovirus. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: OPTN. Target Synonyms: E3-14.7K-interacting proteinFIP-2Huntingtin yeast partner LHuntingtin-interacting protein 7. Accession Number: Q96CV9. Expression Region: 1~577aa. Tag Info: C-Terminal 9Xhis-Tagged. Theoretical MW: 67.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.