Learn More
Biomatik Corporation Recombinant Escherichia coli Acidic protein msyB (msyB), 100UG
Supplier:  Biomatik Corporation RPC20673
Encompass_Preferred
Recombinant Escherichia coli Acidic protein msyB (msyB) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Baculovirus. Endotoxin Level: Not Tested. Species: Escherichia coli (strain K12). Target Name: msyB. Target Synonyms: msyB; b1051; JW1039; Acidic protein MsyB. Accession Number: P25738. Expression Region: 1~124aa. Tag Info: C-Terminal 6Xhis-Tagged. Theoretical MW: 16.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4929