Learn More
Biomatik Corporation Recombinant Mycobacterium bovis 14 kDa antigen (hspX), 100UG
Supplier:  Biomatik Corporation RPC20675
Encompass_Preferred
Recombinant Mycobacterium bovis Alpha-crystallin (hspX) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Baculovirus. Endotoxin Level: Not Tested. Species: Mycobacterium bovis (strain ATCC BAA-935 / AF2122/97). Target Name: hspX. Target Synonyms: 16 kDa antigenHSP 16.3. Accession Number: P0A5B8. Expression Region: 2~144aa. Tag Info: C-Terminal 9Xhis-Tagged. Theoretical MW: 18.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4930