Learn More
Biomatik Corporation Recombinant Human Glutaminyl-peptide cyclotransferase (QPCT), 100UG
Catalog No. 501504570
Encompass_Preferred
Recombinant Human Glutaminyl-peptide cyclotransferase (QPCT) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: QPCT. Target Synonyms: Glutaminyl cyclase; QC; sQCGlutaminyl-tRNA cyclotransferaseGlutamyl cyclase; EC. Accession Number: Q16769. Expression Region: 29~361aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 53.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4570 Supplier Biomatik Corporation Supplier No. RPC20069
Recombinant Human Glutaminyl-peptide cyclotransferase (QPCT) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: QPCT. Target Synonyms: Glutaminyl cyclase; QC; sQCGlutaminyl-tRNA cyclotransferaseGlutamyl cyclase; EC. Accession Number: Q16769. Expression Region: 29~361aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 53.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.