Learn More
Biomatik Corporation Recombinant Mouse Disintegrin and metalloproteinase domain-containing protein 12 (Adam12), partial, 100UG
Supplier:  Biomatik Corporation RPC20690
Encompass_Preferred
Recombinant Mouse Disintegrin and metalloproteinase domain-containing protein 12 (Adam12), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Baculovirus. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Adam12. Target Synonyms: Meltrin-alpha. Accession Number: Q61824. Expression Region: 206~706aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 58.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4932