Learn More
Biomatik Corporation Recombinant Mouse [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 1, mitochondrial (Pdk1), 100UG
Supplier:  Biomatik Corporation RPC20694
Encompass_Preferred
Recombinant Mouse [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 1, mitochondrial (Pdk1) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Baculovirus. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Pdk1. Target Synonyms: Pyruvate dehydrogenase kinase isoform 1. Accession Number: Q8BFP9. Expression Region: 27~434aa. Tag Info: N-Terminal 10Xhis-Tagged. Theoretical MW: 48.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4933