Learn More
Biomatik Corporation Recombinant Human DNA-directed DNA/RNA polymerase mu (POLM), 100UG
Supplier:  Biomatik Corporation RPC20704
Encompass_Preferred
Recombinant Human DNA-directed DNA/RNA polymerase mu (POLM) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Baculovirus. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: POLM. Target Synonyms: Terminal transferase. Accession Number: Q9NP87. Expression Region: 1~494aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 58.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4934