Learn More
Biomatik Corporation Recombinant Human Annexin A4 (ANXA4), 100UG
Supplier:  Biomatik Corporation RPC20904
Encompass_Preferred
Recombinant Human Annexin A4 (ANXA4) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ANXA4. Target Synonyms: 35-beta calcimedinAnnexin IVAnnexin-4Carbohydrate-binding protein p33/p41Chromobindin-4Endonexin ILipocortin IVP32.5PP4-XPlacental anticoagulant protein II. Accession Number: P09525. Expression Region: 2~319aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 55.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4948