Learn More
Biomatik Corporation Recombinant Rabbit Apolipoprotein E (APOE), partial, 100UG
Supplier:  Biomatik Corporation RPC20920
Encompass_Preferred
Recombinant Rabbit Apolipoprotein E (APOE), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rabbit (Oryctolagus cuniculus). Target Name: APOE. Target Synonyms: APOEApolipoprotein E; Apo-E. Accession Number: P18287. Expression Region: 20~311aa. Tag Info: N-Terminal 10Xhis-B2M-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 50.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4952