Learn More
Biomatik Corporation Recombinant Human ATP synthase subunit beta, mitochondrial (ATP5B), partial, 100UG
Catalog No. 501504957
Encompass_Preferred
Recombinant Human ATP synthase subunit beta, mitochondrial (ATP5F1B), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ATP5F1B. Target Synonyms: ATPMB, ATPSB. Accession Number: P06576. Expression Region: 230~529aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 52.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4957 Supplier Biomatik Corporation Supplier No. RPC20955
Recombinant Human ATP synthase subunit beta, mitochondrial (ATP5F1B), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ATP5F1B. Target Synonyms: ATPMB, ATPSB. Accession Number: P06576. Expression Region: 230~529aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 52.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.