Learn More
Biomatik Corporation Recombinant Human UV excision repair protein RAD23 homolog A (RAD23A), 100UG
Supplier:  Biomatik Corporation RPC20071
Encompass_Preferred
Recombinant Human UV excision repair protein RAD23 homolog A (RAD23A) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: RAD23A. Target Synonyms: hHR 23A; hHR23A; HR23A; MGC111083; RAD 23a; RAD23 homolog A (S. cerevisiae); RAD23 homolog A; RAD23 yeast homolog A; RAD23A; RD23A_HUMAN; UV excision repair protein RAD23; UV excision repair protein RAD23 homolog A. Accession Number: P54725. Expression Region: 1~363aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 66.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4572