Learn More
Biomatik Corporation Recombinant Human Neuronal acetylcholine receptor subunit alpha-3 (CHRNA3), partial, 100UG
Catalog No. 501504974
Encompass_Preferred
Recombinant Human Neuronal acetylcholine receptor subunit alpha-3 (CHRNA3), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CHRNA3. Target Synonyms: ACHA3_HUMAN; AChR; Cholinergic receptor neuronal nicotinic alpha polypeptide 3; Cholinergic receptor nicotinic alpha 3; Cholinergic receptor nicotinic alpha polypeptide 3; CHRNA 3; CHRNA3; LNCR2; MGC104879; NACHRA 3. Accession Number: P32297. Expression Region: 32~240aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 44.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4974 Supplier Biomatik Corporation Supplier No. RPC21098
Recombinant Human Neuronal acetylcholine receptor subunit alpha-3 (CHRNA3), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CHRNA3. Target Synonyms: ACHA3_HUMAN; AChR; Cholinergic receptor neuronal nicotinic alpha polypeptide 3; Cholinergic receptor nicotinic alpha 3; Cholinergic receptor nicotinic alpha polypeptide 3; CHRNA 3; CHRNA3; LNCR2; MGC104879; NACHRA 3. Accession Number: P32297. Expression Region: 32~240aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 44.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.