Learn More
Biomatik Corporation Recombinant Mouse Collagen alpha-2 (IV) chain (Col4a2), partial, 100UG
Catalog No. 501504976
Encompass_Preferred
Recombinant Mouse Collagen alpha-2 (IV) chain (Col4a2), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Col4a2. Target Synonyms: Col4a2Collagen alpha-2(IV) chain [Cleaved into: Canstatin]. Accession Number: P08122. Expression Region: 1481~1707aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 29.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4976 Supplier Biomatik Corporation Supplier No. RPC21121
Recombinant Mouse Collagen alpha-2 (IV) chain (Col4a2), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Col4a2. Target Synonyms: Col4a2Collagen alpha-2(IV) chain [Cleaved into: Canstatin]. Accession Number: P08122. Expression Region: 1481~1707aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 29.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.