Learn More
Biomatik Corporation Recombinant Human Protein CYR61 (CYR61), 100UG
Supplier:  Biomatik Corporation RPC21185
Encompass_Preferred
Recombinant Human CCN family member 1 (CCN1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CCN1. Target Synonyms: CCN family member 1Cysteine-rich angiogenic inducer 61Insulin-like growth factor-binding protein 10IBP-10IGF-binding protein 10IGFBP-10Protein GIG1. Accession Number: O00622. Expression Region: 25~381aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 55.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4981