Learn More
Biomatik Corporation Recombinant Human Dickkopf-related protein 1 (DKK1), partial, 100UG
Catalog No. 501504985
Encompass_Preferred
Recombinant Human Dickkopf-related protein 1 (DKK1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: DKK1. Target Synonyms: SK. Accession Number: O94907. Expression Region: 33~266aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 29.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4985 Supplier Biomatik Corporation Supplier No. RPC21213
Recombinant Human Dickkopf-related protein 1 (DKK1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: DKK1. Target Synonyms: SK. Accession Number: O94907. Expression Region: 33~266aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 29.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.