Learn More
Biomatik Corporation Recombinant Human Elongation factor 1-alpha 1 (EEF1A1), 100UG
Supplier:  Biomatik Corporation RPC21238
Encompass_Preferred
Recombinant Human Elongation factor 1-alpha 1 (EEF1A1) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: EEF1A1. Target Synonyms: Elongation factor TuEukaryotic elongation factor 1 A-1Leukocyte receptor cluster member 7EEF1A, EF1A, LENG7. Accession Number: P68104. Expression Region: 1~462aa. Tag Info: Tag-Free. Theoretical MW: 50.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4987