Learn More
Biomatik Corporation Recombinant Human Eukaryotic translation initiation factor 4 gamma 1 (EIF4G1), partial, 100UG
Supplier:  Biomatik Corporation RPC21255
Encompass_Preferred
Recombinant Human Eukaryotic translation initiation factor 4 gamma 1 (EIF4G1), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: EIF4G1. Target Synonyms: p220. Accession Number: Q04637. Expression Region: 1250~1599aa. Tag Info: N-Terminal 10Xhis-B2M-Jd-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 46.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4988