Learn More
Biomatik Corporation Recombinant Human Nuclear receptor ROR-gamma (RORC), 100UG
Supplier:  Biomatik Corporation RPC20074
Encompass_Preferred
Recombinant Human Nuclear receptor ROR-gamma (RORC) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: RORC. Target Synonyms: Nuclear receptor RZR-gammaNuclear receptor subfamily 1 group F member 3RAR-related orphan receptor CRetinoid-related orphan receptor-gamma. Accession Number: P51449. Expression Region: 1~518aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 74.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4575