Learn More
Biomatik Corporation Recombinant Mouse Alpha-enolase (Eno1), partial, 100UG
Catalog No. 501504989
Encompass_Preferred
Recombinant Mouse Alpha-enolase (Eno1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Eno1. Target Synonyms: 2-phospho-D-glycerate hydro-lyaseEnolase 1Non-neural enolase. Accession Number: P17182. Expression Region: 2~433aa. Tag Info: N-Terminal 10Xhis-Tagged. Theoretical MW: 50.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4989 Supplier Biomatik Corporation Supplier No. RPC21262
Recombinant Mouse Alpha-enolase (Eno1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Eno1. Target Synonyms: 2-phospho-D-glycerate hydro-lyaseEnolase 1Non-neural enolase. Accession Number: P17182. Expression Region: 2~433aa. Tag Info: N-Terminal 10Xhis-Tagged. Theoretical MW: 50.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.